Learn More
Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II), Pd 16.3%, Thermo Scientific Chemicals
Description
Coupling reactions, Suzuki couplings.Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene] palladium(II) is used as a catalyst in Suzuki-cross coupling reactions, which involve reaction of aromatic halides with methyliminodiacetic acid derivatives. it is also used as a catalyst for the preparation of chalcones. It is used in the preparation of (bisphosphinometallocene)-palladium(II) chloride complexes, which used as a starting material for the Buchwald-Hartwig reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Specifications
Color | Red |
Quantity | 0.1 g |
Solubility Information | Soluble in dichloromethane and chloroform. Partially soluble in acetone,alcohols,tertrahydrofuran and dioxane. |
Formula Weight | 651.77 |
Concentration or Composition (by Analyte or Components) | Pd 16.3% |
Physical Form | Crystalline |
Chemical Name or Material | Dichloro[1,1'-bis (di-tert-butylphosphino)ferrocene]palladium (II) |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.