Learn More
Dichloro[1,2-bis(diphenylphosphino)ethane]palladium(II), Pd 18.5%, Thermo Scientific™
£79.26 - £281.00
Chemical Identifiers
CAS | 19978-61-1 |
---|---|
Molecular Formula | C26H26Cl2P2Pd+2 |
Molecular Weight (g/mol) | 577.762 |
MDL Number | MFCD00015702 |
InChI Key | LDJXFZUGZASGIW-UHFFFAOYSA-N |
Synonym | dichloro 1,2-bis diphenylphosphino ethane palladium ii |
PubChem CID | 131675886 |
IUPAC Name | 2-diphenylphosphanylethyl(diphenyl)phosphane;palladium(2+);dihydrochloride |
SMILES | C1=CC=C(C=C1)P(CCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl.[Pd+2] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
19978-61-1 | |
577.762 | |
LDJXFZUGZASGIW-UHFFFAOYSA-N | |
131675886 | |
C1=CC=C(C=C1)P(CCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl.[Pd+2]Show More |
C26H26Cl2P2Pd+2 | |
MFCD00015702 | |
dichloro 1,2-bis diphenylphosphino ethane palladium ii | |
2-diphenylphosphanylethyl(diphenyl)phosphane;palladium(2+);dihydrochloride |
Specifications
19978-61-1 | |
Yellow | |
PdCl2[(C6H5)2PCH2CH2P(C6H5)2] | |
Hygroscopic | |
LDJXFZUGZASGIW-UHFFFAOYSA-N | |
2-diphenylphosphanylethyl(diphenyl)phosphane;palladium(2+);dihydrochloride | |
131675886 | |
Pd 18.5% | |
Dichloro[1,2-bis(diphenylphosphino)ethane]palladium(II) |
>360°C | |
C26H26Cl2P2Pd+2 | |
MFCD00015702 | |
dichloro 1,2-bis diphenylphosphino ethane palladium ii | |
C1=CC=C(C=C1)P(CCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl.[Pd+2] | |
577.762 | |
575.75 | |
Powder |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
Your input is important to us. Please complete this form to provide feedback related to the content on this product.