missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II), Pd 12.1%, Thermo Scientific Chemicals

£148.74 - £585.92

Chemical Identifiers

CAS 205319-10-4
Molecular Formula C39H32Cl2OP2Pd
Molecular Weight (g/mol) 755.95
MDL Number MFCD14155707
InChI Key MNBAPNBMBOYNAW-UHFFFAOYSA-L
Synonym dichloro 9,9-dimethyl-4,5-bis diphenylphosphino xanthene palladium ii, palladium 2+ xantphos dichloride, 4,5-bis diphenylphosphino-9,9-dimethylxanthene dichloropalladium ii, 4,5-bis diphenylphosphino-9,9-dimethylxanthene palladium ii dichloride
PubChem CID 86280406
IUPAC Name (5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl)-diphenylphosphane;palladium(2+);dichloride
SMILES Cl[Pd++]Cl.CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15465025
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
046665.03
1 g
£148.74
1g
Please to purchase this item. Need a web account? Register with us today!
15475025
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
046665.06
5 g
£585.92
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

205319-10-4
755.95
MNBAPNBMBOYNAW-UHFFFAOYSA-L
86280406
Cl[Pd++]Cl.CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
C39H32Cl2OP2Pd
MFCD14155707
dichloro 9,9-dimethyl-4,5-bis diphenylphosphino xanthene palladium ii, palladium 2+ xantphos dichloride, 4,5-bis diphenylphosphino-9,9-dimethylxanthene dichloropalladium ii, 4,5-bis diphenylphosphino-9,9-dimethylxanthene palladium ii dichloride
(5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl)-diphenylphosphane;palladium(2+);dichloride

Specifications

205319-10-4
Yellow
MFCD14155707
MNBAPNBMBOYNAW-UHFFFAOYSA-L
(5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl)-diphenylphosphane;palladium(2+);dichloride
86280406
Pd 14.1%
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II)
280°C to 287°C (decomposition)
C39H32Cl2OP2Pd
dichloro 9,9-dimethyl-4,5-bis diphenylphosphino xanthene palladium ii, palladium 2+ xantphos dichloride, 4,5-bis diphenylphosphino-9,9-dimethylxanthene dichloropalladium ii, 4,5-bis diphenylphosphino-9,9-dimethylxanthene palladium ii dichloride
Cl[Pd++]Cl.CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
755.95
755.96
Crystalline Solid
Videos
Safety and Handling

Safety and Handling

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

missing translation for 'einecsNumber' : 685-312-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.