missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloro[bis(1,3-diphenylphosphino)propane]palladium(II), Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
(1,3-Bis(diphenylphosphino)propane)palladium(II) chloride, used ubiquitously as a employed preparation of palladium for cross-coupling protocols in organic synthesis. It is also a useful organo-metallic chemical catalyst. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(1,3-Bis(diphenylphosphino)propane)palladium(II) chloride, used ubiquitously as a employed preparation of palladium for cross-coupling protocols in organic synthesis. It is also a useful organo-metallic chemical catalyst. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in coupling reactions.
Solubility
Soluble in dichloromethane.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Air Sensitive.
(1,3-Bis(diphenylphosphino)propane)palladium(II) chloride, used ubiquitously as a employed preparation of palladium for cross-coupling protocols in organic synthesis. It is also a useful organo-metallic chemical catalyst. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in coupling reactions.
Solubility
Soluble in dichloromethane.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Air Sensitive.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 g |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in dichloromethane. |
Formula Weight | 589.77 |
Chemical Name or Material | Dichloro[bis(1,3-diphenylphosphino)propane]palladium(II) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'