missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II), Pd 17.6%, Thermo Scientific Chemicals

£171.86 - £728.63

Chemical Identifiers

CAS 29964-62-3
Molecular Formula C28H28Cl2P2Pd
Molecular Weight (g/mol) 603.80
MDL Number MFCD02093437
InChI Key JQXJBXVWVPVTOO-UHFFFAOYSA-L
Synonym 1,4-bis diphenylphosphino butane-palladium ii chloride, 1,4-butylenebis diphenylphosphine palladium dichloride, pdcl2 dppb, 1,4-bis diphenylphosphino butane palladium ii dichloride, palladium ii chloride-1,4-bis diphenylphosphine butane, dichloro bis 1,4-diphenylphosphino butane palladium ii, palladium ii chloride 1,4-bis diphenylphosphino butane complex, dichloro 1,4-bis diphenylphosphino butane palladium ii, pd dppb cl2, acmc-209sk1
PubChem CID 10974057
IUPAC Name dichloropalladium;4-diphenylphosphanylbutyl(diphenyl)phosphane
SMILES Cl[Pd]Cl.C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15436655
View Documents
Thermo Scientific Alfa Aesar
044971.03
1 g
£171.86
1g
Please to purchase this item. Need a web account? Register with us today!
15446655
View Documents
Thermo Scientific Alfa Aesar
5 g
£728.63
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II) is used for the catalysis of styrene carbonylation, coupling of alkyl Grignard reagents with organic halides, selective monoalkylation of organic polyhalides, and modification of the dihalovinyl moiety of synthetic pyrethroids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

29964-62-3
603.80
JQXJBXVWVPVTOO-UHFFFAOYSA-L
10974057
Cl[Pd]Cl.C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
C28H28Cl2P2Pd
MFCD02093437
1,4-bis diphenylphosphino butane-palladium ii chloride, 1,4-butylenebis diphenylphosphine palladium dichloride, pdcl2 dppb, 1,4-bis diphenylphosphino butane palladium ii dichloride, palladium ii chloride-1,4-bis diphenylphosphine butane, dichloro bis 1,4-diphenylphosphino butane palladium ii, palladium ii chloride 1,4-bis diphenylphosphino butane complex, dichloro 1,4-bis diphenylphosphino butane palladium ii, pd dppb cl2, acmc-209sk1
dichloropalladium;4-diphenylphosphanylbutyl(diphenyl)phosphane

Specifications

29964-62-3
MFCD02093437
Soluble in ethanol,benzene,and ethereal solvents.
Cl[Pd]Cl.C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
603.80
603.8
Lumps
C28H28Cl2P2Pd
1,4-bis diphenylphosphino butane-palladium ii chloride, 1,4-butylenebis diphenylphosphine palladium dichloride, pdcl2 dppb, 1,4-bis diphenylphosphino butane palladium ii dichloride, palladium ii chloride-1,4-bis diphenylphosphine butane, dichloro bis 1,4-diphenylphosphino butane palladium ii, palladium ii chloride 1,4-bis diphenylphosphino butane complex, dichloro 1,4-bis diphenylphosphino butane palladium ii, pd dppb cl2, acmc-209sk1
JQXJBXVWVPVTOO-UHFFFAOYSA-L
dichloropalladium;4-diphenylphosphanylbutyl(diphenyl)phosphane
10974057
Pd 17.6%
Dichloro[bis (1,4-diphenylphosphino)butane]palladium (II)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II)

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II)

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only