missing translation for 'onlineSavingsMsg'
Learn More
Dichloroacetic acid, 99+%, Thermo Scientific Chemicals Product Code.: 10500551

Dichloroacetic acid, 99+%, Thermo Scientific Chemicals

Product Code. 10500551
250 mL, Glass bottle
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10500551

Brand: Thermo Scientific Acros 113062500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-43-6
Molecular Formula C2H2Cl2O2
Molecular Weight (g/mol) 128.94
MDL Number MFCD00004223
InChI Key JXTHNDFMNIQAHM-UHFFFAOYSA-N
Synonym dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid
PubChem CID 6597
ChEBI CHEBI:36386
IUPAC Name 2,2-dichloroacetic acid
SMILES C(C(=O)O)(Cl)Cl

Specifications

Melting Point 9.0°C to 11.0°C
Color Brown
Density 1.5600g/mL
Boiling Point 194.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula Cl2CHCO2H
Refractive Index 1.4653 to 1.4673
Quantity 250 mL
Beilstein 02,202
Fieser 16,115
Merck Index 15,3059
Specific Gravity 1.56
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol,ether and ethanol,soluble in acetone
Formula Weight 128.94
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Dichloroacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichloroacetic acid
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichloroacetic acid, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.