missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(tri-o-tolylphosphine)palladium(II), 95%, Thermo Scientific Chemicals

£48.04 - £211.05

Chemical Identifiers

CAS 40691-33-6
Molecular Formula C42H42Cl2P2Pd
Molecular Weight (g/mol) 786.06
MDL Number MFCD00274659
InChI Key GMLMZLGOMMIIMA-UHFFFAOYSA-L
Synonym palladium,dichlorobis tris 2-methylphenyl phosphine
PubChem CID 90656943
IUPAC Name palladium;tris(2-methylphenyl)phosphane;dichloride
SMILES Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15456655
View Documents
Thermo Scientific Alfa Aesar
044976.03
1 g
£48.04
1g
Please to purchase this item. Need a web account? Register with us today!
15466655
View Documents
Thermo Scientific Alfa Aesar
044976.06
5 g
£211.05
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dichlorobis(tri-o-tolylphosphine)palladium(II) is used in reaction of tributyltin enolates, prepared in situ from tributyltin methoxide and enol acetates, with aryl bromides, coupling reaction of aryl bromides with vinylic acetates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

40691-33-6
786.06
GMLMZLGOMMIIMA-UHFFFAOYSA-L
90656943
Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1
C42H42Cl2P2Pd
MFCD00274659
palladium,dichlorobis tris 2-methylphenyl phosphine
palladium;tris(2-methylphenyl)phosphane;dichloride

Specifications

40691-33-6
C42H42Cl2P2Pd
Air and moisture sensitive
Soluble in ethanol,benzene,and ethereal solvents.
Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1
786.06
786.08
Powder
Yellow
MFCD00274659
palladium,dichlorobis tris 2-methylphenyl phosphine
GMLMZLGOMMIIMA-UHFFFAOYSA-L
palladium;tris(2-methylphenyl)phosphane;dichloride
90656943
98%
Dichlorobis (tri-o-tolylphosphine)palladium (II)
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only