missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodiphenylsilane, 97%, Thermo Scientific Chemicals

Product Code. 10080900
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-10-4
Molecular Formula C12H10Cl2Si
Molecular Weight (g/mol) 253.20
MDL Number MFCD00000489
InChI Key OSXYHAQZDCICNX-UHFFFAOYSA-N
Synonym diphenyldichlorosilane, silane, dichlorodiphenyl, dichloro diphenyl silane, diphenylsilicon dichloride, diphenylsilyl dichloride, dichlor-difenylsilan, diphenyl dichlorosilane, dichlor-difenylsilan czech, hsdb 316, benzene, 1,1'-dichlorosilylene bis
PubChem CID 6627
IUPAC Name dichloro(diphenyl)silane
SMILES Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point -22.0°C
Color Brown or Colorless to Yellow
Density 1.2040g/mL
Boiling Point 305.0°C
Flash Point 155°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2SiCl2
Refractive Index 1.5765 to 1.5815
Quantity 100 mL
Beilstein 16,91
Fieser 13,74
Specific Gravity 1.204
Solubility Information Solubility in water: decomposes
Formula Weight 253.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dichlorodiphenylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichlorodiphenylsilane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'