Learn More
Dichloroisocyanuric acid sodium salt, 98%, Thermo Scientific Chemicals
Dichloroisocyanuric acid sodium salt, >-97.5% (on dry substance), C3HCl2N3NaO3, CAS Number-2893-78-9, sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt, 500g, 98%, White, 220-767-7, 219.95, Danger
£24.22 - £122.61
Chemical Identifiers
CAS | 2893-78-9 |
---|---|
Molecular Formula | C3HCl2N3NaO3 |
Molecular Weight (g/mol) | 220.949 |
MDL Number | MFCD00006036 |
InChI Key | UNWRHVZXVVTASG-UHFFFAOYSA-N |
Synonym | sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt |
PubChem CID | 86657659 |
IUPAC Name | 1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium |
SMILES | C1(=O)NC(=O)N(C(=O)N1Cl)Cl.[Na] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10712251
|
Thermo Scientific Acros
204110250 |
25 g |
£24.22
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10451631
|
Thermo Scientific Acros
204115000 |
500 g |
£37.79
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15274674
|
Thermo Scientific Acros
204110025 |
2.5 kg |
£122.61
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagentChemical Identifiers
2893-78-9 | |
220.949 | |
UNWRHVZXVVTASG-UHFFFAOYSA-N | |
86657659 | |
C1(=O)NC(=O)N(C(=O)N1Cl)Cl.[Na] |
C3HCl2N3NaO3 | |
MFCD00006036 | |
sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt | |
1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium |
Specifications
2893-78-9 | |
6 to 7 (1% aq. soln. at 25°C) | |
98% | |
C3HCl2N3NaO3 | |
sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt | |
C1(=O)NC(=O)N(C(=O)N1Cl)Cl.[Na] | |
220.949 | |
219.95 | |
Crystals or Granules |
White | |
Authentic | |
Plastic bottle | |
MFCD00006036 | |
UNWRHVZXVVTASG-UHFFFAOYSA-N | |
1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium | |
86657659 | |
≥97.5% (on dry substance) | |
Dichloroisocyanuric acid sodium salt, 98% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life.
Very toxic to aquatic life with long lasting effects.
May intensify fire; oxidizer.
Contact with acids liberates toxic gas.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 220-767-7
missing translation for 'rtecsNumber' : XZ1900000
missing translation for 'tsca' : TSCA
- Dichloroisocyanuric acid, sodium salt
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only