missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, 99.5%, for analysis, stabilized with ethanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | -97.0°C |
Acidity | 0.0005 meq/g max. |
Density | 1.3250g/mL |
Boiling Point | 39.0°C to 40.0°C |
Quantity | 2.5 L |
Packaging | Glass Bottle |
Physical Form | Liquid |
Refractive Index | 1.4230 to 1.4250 |
Assay Percent Range | 99.4% min. (GC) |
Residue after Evaporation | 0.0005% max. |
Show More |
Safety and Handling
Product Identifier
- Dichloromethane, stabilized with ethanol
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'