missing translation for 'onlineSavingsMsg'
Learn More
Dichloromethane, 99.8+%, for analysis, stabilized with amylene, Thermo Scientific Chemicals Product Code.: 10162330

Dichloromethane, 99.8+%, for analysis, stabilized with amylene, Thermo Scientific Chemicals

Product Code. 10162330
25 L, Metal Drum
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Glass Bottle
Metal Drum
Plastic Drum
This item is not returnable. View return policy

Product Code. 10162330

Brand: Thermo Scientific Acros 113460250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2, 513-35-9
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97.0°C
Density 1.3250g/mL
Boiling Point 39.0°C to 40.0°C
Quantity 25 L
Packaging Metal Drum
Physical Form Liquid
Assay Percent Range 99.8+%
Residue after Evaporation 0.001% max.
Specific Gravity 1.325
Linear Formula CH2Cl2
Viscosity 0.43 mPa.s (20°C)
Beilstein 01, 60
Water 0.02% max. (Coulometric)
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Merck Index 15, 6135
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Formula Weight 84.93
Percent Purity ≥99.8%
Grade Analytical
Chemical Name or Material Dichloromethane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2534176 Certificate TypeCertificate of Analysis Date09/04/2025 Catalog Number113460025,113460010,113460000,113460100,113460050,113460051,113460250
Lot Number1913328 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number113460025,113460010,113460100,113460050,113460250
Lot Number1996119 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number113460025,113460010,113460100,113460050,113460250
Lot Number2054350 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number113460025,113460010,113460100,113460050,113460250
Lot Number1900558 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number113460025,113460010,113460100,113460050,113460250
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichloromethane, 99.8+%, for analysis, stabilized with amylene, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.