missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, 99.8+%, for analysis, stabilized with amylene, Thermo Scientific™

Product Code. 10771545
Click to view available options
5L,Tin Can
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -97°C
CAS 75-09-2
Color Colorless
Appearance Clear liquid
Decomposition Information Decomposition temperature: >120°C
Density 1.325
Boiling Point 39°C to 40°C (760 mmHg)
Quantity 5 L
Identification Passes test
Ignition Point 556°C (auto-ignition)
Packaging Tin Can
Physical Form Liquid
Residue after Evaporation =<0.001 %
Molecular Formula CH2Cl2
Linear Formula CH2Cl2
MDL Number MFCD00000881
Vapor Pressure 350 Mbar (20°C)
UN Number 1593
Viscosity 0.43 mPa/s (20°C)
Water =<0.02% (Coulometric)
Merck Index 15, 6135
Synonym Methylene chloride
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES ClCCl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.93
Formula Weight 84.93
Percent Purity 99.8+%
Grade Analytical
Chemical Name or Material Dichloromethane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'