missing translation for 'onlineSavingsMsg'
Learn More
Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals Product Code.: 10046082

Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals

Product Code. 10046082
5 L
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
This item is not returnable. View return policy

Product Code. 10046082

Brand: Thermo Scientific Acros 383780050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Density 1.325g/cm3
Quantity 5 L
Packaging Plastic Drum
Assay Percent Range 99+%
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Formula Weight 84.93
Chemical Name or Material Dichloromethane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2534176 Certificate TypeCertificate of Analysis Date09/04/2025 Catalog Number383780025,383780010,383780250,383780050
Lot Number2523247 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number383780025,383780010,383780250,383780050
Lot Number2412238 Certificate TypeCertificate of Analysis Date23/12/2024 Catalog Number383780025,383780010,383780250,383780050
Lot Number2406292 Certificate TypeCertificate of Analysis Date23/12/2024 Catalog Number383780025,383780010,383780250,383780050
Lot Number2496383 Certificate TypeCertificate of Analysis Date23/12/2024 Catalog Number383780025,383780010,383780250,383780050
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.