missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, ACS Reagent, contains 40-150 amylene as stabilizer, ≥99.5%, Honeywell Riedel-de Haën™

Catalog Number 15697830
Click to view available options
Quantity:
1 L
2.5 L
10 L
20 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
75-09-2
CH2Cl2
84.93
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
6344
CHEBI:15767
dichloromethane
ClCCl
This item is not returnable. View return policy
75-09-2
CH2Cl2
84.93
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
6344
CHEBI:15767
dichloromethane
ClCCl

ACS Reagent, contains 40-150 amylene as stabilizer,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Melting Point -97°C (literature)
CAS Min % ≥99.0000%
CAS Max % ≥99.0000%
Linear Formula CH2Cl2
UN Number UN1593
Beilstein 1730800
Formula Weight 84.93
Percent Purity >99.5%
Grade ACS Reagent
Density 1.325 g/mL at 25 °C(lit.)
Boiling Point 39°C to 41°C
Quantity 20 L
Packaging Stainless Steel Can
Water max. 0.02%
Chemical Name or Material Dichloromethane, ACS Reagent, contains 40-150 amylene as stabilizer, ≥99.5%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'