missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic Canister
Plastic-coated Glass Bottle
Description
CH2Cl2, CAS Number-75-09-2, aerothene mm, methane, dichloro, methane dichloride, methylene bichloride, methylene chloride, methylene dichloride, solaesthin, freon 30, narkotil, solmethine, 2.5L, 39 deg.C, CHEBI:15767, Colorless, 84.93g/mol, YMWUJEATGCHHMB-UHFFFAOYSA-N, dichloromethane, -97 deg.C

Specifications
Specifications
Melting Point | -97°C |
Color | Colorless |
Boiling Point | 39°C |
Quantity | 1 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 350mbar at 20°C |
Viscosity | 0.43 mPaS at 20°C |
Formula Weight | 84.93g/mol |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2526194 | Certificate of Analysis | 07/02/2025 | D/1852/PB15X,D/1852/17,D/1852/17X,D/1852/27,D/1852/15,D/1852/17Z,D/1852/25,D/1852/24,D/1852/15Z,D/1852/PB17,D/1852/PB15,D/1852/21,D/1852/PC15 |
Safety and Handling


Product Identifier
- Dichloromethane
- Warning
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H336-May cause drowsiness or dizziness.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction