missing translation for 'onlineSavingsMsg'
Learn More
Riedel-de Haën Dichloromethane CHROMASOLV™ Product Code.: 15995480
compliance-icons compliance-icons

Riedel-de Haën Dichloromethane CHROMASOLV™

Product Code. 15995480
7 L
Click to view available options
Quantity:
7 L
45 L
This item is not returnable. View return policy

Product Code. 15995480

Brand: Riedel-de Haën 344887L

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Dichloromethane CHROMASOLV™ for pesticide residue analysis, ≥99.8% (GC).

  • For gas chromatography (GC).
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC.
  • Non-volatile matter and water content are specified for each solvent.

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym Methylene Chloride, DCM
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

pH Not applicable
Melting Point -97°C
CAS Min % ≥99.00%
Color Colorless
Density 1.32 to 1.33 g/cm3 (20°C)
Boiling Point 39°C to 41°C (1.013 hPa)
Quantity 7 L
Flash Point Not applicable
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line CHROMASOLV™
Linear Formula CH2Cl2
Vapor Pressure 453 hPa (20°C)
UN Number UN1593
Viscosity 0.44 mPa.s (20°C)
Beilstein 1730800
Percent Purity ≥99.80%
Grade GC
Chemical Name or Material Dichloromethane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Riedel-de Haën Dichloromethane CHROMASOLV™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.