missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane-d{2}, 99.9% (Isotopic), Thermo Scientific Chemicals

Product Code. 11357958
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

In Anion-templated assembly of [2] rotaxanes, the addition of tetrabutylammonium (TBA) halide, dihydrogen phosphate and hydrogen sulfate salts to dichloromethane-d 2 solutions of both derivatives resulted in significant downfield shift perturbations of the respective compounds' aryl b and amide c protons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1665-00-5
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 86.939
MDL Number MFCD00000882
InChI Key YMWUJEATGCHHMB-DICFDUPASA-N
Synonym dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
PubChem CID 160586
IUPAC Name dichloro(dideuterio)methane
SMILES C(Cl)Cl

Specifications

Physical Form Liquid
Density 1.35 g/mL
Odor Sweet
Sensitivity Moisture
Quantity 1 g
UN Number UN1593
Solubility Information Insoluble in water. Soluble in acetone,chloroform,tetrachloromethane,dimethylfomrarnide.
Formula Weight 86.95
Percent Purity 99.9%
Chemical Name or Material Dichloromethane-d2, Isotopic

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'