missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane-d2, for NMR, 99.5 atom % D, AcroSeal™, Thermo Scientific Chemicals

Product Code. 11944371
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Spécification

CAS 1665-00-5
Melting Point -97.0°C
Color Colorless
Density 1.3600g/mL
Boiling Point 40.0°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula CCl2D2
Linear Formula CD2Cl2
Refractive Index 1.4218
Quantity 100 mL
Specific Gravity 1.36
Synonym dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
Solubility Information Solubility in water: .. Other solubilities: soluble in alchol,ether and dmf
InChI Key YMWUJEATGCHHMB-DICFDUPASA-N
SMILES C(Cl)Cl
IUPAC Name dichloro(dideuterio)methane
Molecular Weight (g/mol) 86.95
PubChem CID 160586
Formula Weight 86.95
Grade NMR
Physical Form Liquid
Chemical Name or Material Dichloromethane-d2, For NMR
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • Dichloromethane-d2
Mention d'avertissement
  • Warning
Catégorie de danger
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Informations supplémentaires
  • MIXTURE LIST-Contains : Dichloro(2H2)methane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'