missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane-d2, for NMR, 99.8 atom% D

Product Code. p-106747 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass Bottle
Unit Size:
10g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10710981

Brand: Thermo Scientific Acros 321270050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1665-00-5 | CH2Cl2 | 86.939 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1665-00-5
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 86.939
MDL Number MFCD00000882
InChI Key YMWUJEATGCHHMB-DICFDUPASA-N
Synonym dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
PubChem CID 160586
IUPAC Name dichloro(dideuterio)methane
SMILES C(Cl)Cl

Specifications

Physical Form Liquid
Color Colorless
Density 1.3600g/mL
Deuterium (D) 99.75% min.
Packaging Glass Bottle
Refractive Index 1.4218
Water 0.02% max. (Coulometric)
Quantity 5 g
Melting Point -97.0°C
Boiling Point 40.0°C
Linear Formula CD2Cl2
Beilstein 01, IV, 39
Formula Weight 86.95
Grade NMR
Chemical Name or Material Dichloromethane-d2, 99.8 atom% D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane-d2
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Dichloromethane-d2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.