missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, Honeywell™
Click to view available options
5L ALUMINUM BOTTLE
Description
- ≥95% for general laboratory use.
Specifications
CAS | 75-09-2 |
PubChem CID | 6344 |
ChEBI | CHEBI:15767 |
Percent Purity | min. 99.9 Gew.% |
Grade | Reagent |
Boiling Point | 39°C to 41°C (1.013 hPa) |
Quantity | 5 L |
Packaging | Aluminum Bottle |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'