missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
CAS | 75-09-2 |
---|---|
Molecular Formula | CH2Cl2 |
Molecular Weight (g/mol) | 84.93 |
MDL Number | MFCD00000881 |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
PubChem CID | 6344 |
ChEBI | CHEBI:15767 |
IUPAC Name | dichloromethane |
SMILES | ClCCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15602590
|
Honeywell Riedel-de Haën
24233-1L |
1 L |
£31.04
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15612590
|
Honeywell Riedel-de Haën
24233-2.5L |
2.5 L |
£79.05
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15632590
|
Honeywell Riedel-de Haën
24233-35KG |
35 kg |
£277.81
35kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15622590
|
Honeywell Riedel-de Haën
24233-250KG |
250 kg |
£1240.40
250kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
75-09-2 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
6344 | |
dichloromethane |
CH2Cl2 | |
MFCD00000881 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
CHEBI:15767 | |
ClCCl |
Specifications
75-09-2 | |
0.22 | |
CH2Cl2 | |
24.45psi (55°C), 6.83psi (20°C) | |
1730800 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
6344 | |
84.93g/mol | |
puriss. |
1.318 to 1.322g/mL at 25°C, 1.325g/mL at 25°C | |
1223°F | |
MFCD00000881 | |
1593 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
ClCCl | |
84.93 | |
CHEBI:15767 | |
≥99% (GC) | |
Dichloromethane, Meets analytical specification of Ph.Eur.NF |
Safety and Handling
P261-P281-P305 + P351 + P338
H315-H319-H335-H336-H351-H373
Warning
missing translation for 'einecsNumber' : 200-838-9
missing translation for 'rtecsNumber' : PA8050000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Dichloromethane
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.