missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, Puriss. p.a., ACS Reagent, Reag. ISO, ≥99.9% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
10 L
25 L
35 kg
180 L
250 kg
Specifications
Density | 1.325g/mL at 25°C, 1.320 to 1.330g/Lat 20°C |
Quantity | 1 L |
Explosion Limit | 0.22 |
Ignition Point | 1223°F |
Vapor Pressure | 6.83psi (20°C), 24.45psi (55°C) |
UN Number | 1593 |
Beilstein | 1730800 |
Formula Weight | 84.93g/mol |
Percent Purity | ≥99.9% (GC) |
Grade | ACS/puriss. p.a. |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Dichloromethane
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'