Learn More
(Dichloromethylene)dimethylammonium chloride, 95%, Thermo Scientific Chemicals
£55.30 - £172.00
Chemical Identifiers
CAS | 33842-02-3 |
---|---|
Molecular Formula | C3H6Cl3N |
Molecular Weight (g/mol) | 162.44 |
MDL Number | MFCD00011870 |
InChI Key | NRNFKRFWZQQDMD-UHFFFAOYSA-M |
Synonym | dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride |
PubChem CID | 2723945 |
IUPAC Name | dichloromethylidene(dimethyl)azanium;chloride |
SMILES | [Cl-].CN(C)[C+](Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11405043
|
Thermo Scientific Alfa Aesar
A12300.14 |
25 g |
£55.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11415043
|
Thermo Scientific Alfa Aesar
A12300.22 |
100 g |
£172.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
33842-02-3 | |
162.44 | |
NRNFKRFWZQQDMD-UHFFFAOYSA-M | |
2723945 | |
[Cl-].CN(C)[C+](Cl)Cl |
C3H6Cl3N | |
MFCD00011870 | |
dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride | |
dichloromethylidene(dimethyl)azanium;chloride |
Specifications
33842-02-3 | |
C3H6Cl3N | |
UN3261 | |
Moisture sensitive | |
NRNFKRFWZQQDMD-UHFFFAOYSA-M | |
dichloromethylidene(dimethyl)azanium;chloride | |
2723945 | |
95% |
∼185°C (decomposition) | |
MFCD00011870 | |
4157987 | |
dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride | |
[Cl-].CN(C)[C+](Cl)Cl | |
162.44 | |
162.45 | |
(Dichloromethylene)dimethylammonium chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 251-695-4
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only