missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethylsilane, 97+%

Product Code. 11357687
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11357687

Brand: Thermo Scientific Alfa Aesar 014079.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-54-7

A monomer (‘building block’) in the production of silicone polymers. An intermediate (starting material) in the production of other organic and inorganic chemicals. In the electronics industry for the production of ultra-pure polysilicon and in the manufacture of semiconductors and photovoltaics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-54-7
MDL Number MFCD00000494
Synonym Methyldichlorosilane

Specifications

Boiling Point 41°C to 42°C
Melting Point -93°C
Color Colorless
Physical Form Liquid
Quantity 500 g
Linear Formula CH3SiHCl2
UN Number UN1242
Beilstein 1071194
Flash Point -32°C (-26°F)
Solubility Information Soluble in heptane,benzene,ether.
Formula Weight 115.03
Percent Purity ≥97%
Refractive Index 1.398
Sensitivity Air and moisture sensitive
Density 1.105 g/mL
Chemical Name or Material Dichloromethylsilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.