missing translation for 'onlineSavingsMsg'
Learn More
Dichloromethylvinylsilane, 97%, Thermo Scientific Chemicals Product Code.: 10366690

Dichloromethylvinylsilane, 97%, Thermo Scientific Chemicals

Product Code. 10366690
500 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10366690

Brand: Thermo Scientific Acros 147435000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-70-9
MDL Number MFCD00000492
InChI Key YLJJAVFOBDSYAN-UHFFFAOYSA-N
Synonym dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silaneShow MoreShow Less
PubChem CID 31299
IUPAC Name dichloro-ethenyl-methylsilane
SMILES C[Si](C=C)(Cl)Cl

Specifications

Color Colorless to Brown
Density 1.0800g/mL
Boiling Point 92.0°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C3H6Cl2Si
Linear Formula H2C=CHSi(CH3)Cl2
Refractive Index 1.4290 to 1.4310
Quantity 500 g
Specific Gravity 1.08
Solubility Information Solubility in water: reacts.
Molecular Weight (g/mol) 141.07
Formula Weight 141.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dichloromethylvinylsilane, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0451153 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number147435000
Lot NumberA0402030 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number147430025,147431000,147435000
Lot NumberA0377977 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number147430025,147431000,147435000
Lot NumberA0414515 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number147430025,147431000,147435000
compliance-icons compliance-icons
Product Identifier
  • Dichloromethylvinylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichloromethylvinylsilane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.