missing translation for 'onlineSavingsMsg'
Learn More

Dichlorophenylborane, 96%, Thermo Scientific Chemicals

Product Code. 11378497
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Dichlorophenylborane is widely used as a catalyst in Antiproliferative macrolide and cell migration inhibitor lactimidomycin and in enantioselective synthesis of polypeptide segments using vinylogous Mukaiyama aldol reactions and in enantioselective Diels-Alder cycloadditions after its reaction with allo-threonine derivatives. It is a stereo selective alkylative ring opening of cyclic anhydrides and in cross-metathesis reaction of amino derivatives with olefines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-51-8
Molecular Formula C6H5BCl2
Molecular Weight (g/mol) 158.816
MDL Number MFCD00013612
InChI Key NCQDQONETMHUMY-UHFFFAOYSA-N
Synonym phenylboron dichloride, borane, dichlorophenyl, dichloro phenyl borane, phenyldichloroborane, phenylboron dibromide, dichlorophenylboron, dichloro-phenyl borane, dichlorophenylborane, bis chloranyl-phenyl-borane, dichloro-phenyl-boron
PubChem CID 136678
IUPAC Name dichloro(phenyl)borane
SMILES B(C1=CC=CC=C1)(Cl)Cl

Specifications

Melting Point 7°C
Color Colorless
Density 1.194 g/mL
Boiling Point 175°C
Flash Point 14°C (57°F)
Odor Acrid
Refractive Index 1.545
Quantity 1 g
UN Number UN2924
Beilstein 2243576
Sensitivity Moisture sensitive
Solubility Information Soluble in water (Reacts violently).
Formula Weight 158.83
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Dichlorophenylborane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'