missing translation for 'onlineSavingsMsg'
Learn More
μ-Dichlorotetraethylene dirhodium(I), Thermo Scientific Chemicals Product Code.: 10144120

μ-Dichlorotetraethylene dirhodium(I), Thermo Scientific Chemicals

Product Code. 10144120 Shop All Thermo Scientific Products
100 mg, Glass bottle
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10144120

Brand: Thermo Scientific Acros 197591000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12081-16-2
Molecular Formula C8H16Cl2Rh2
Molecular Weight (g/mol) 388.93
MDL Number MFCD00013206
InChI Key OOQJCPOXJFCGCR-UHFFFAOYSA-L
Synonym chlorobis ethylene rhodium i dimer, dichlorotetra ethylene dirhodium i
PubChem CID 45357812
IUPAC Name ethene;rhodium;dichloride
SMILES Cl[Rh].Cl[Rh].C=C.C=C.C=C.C=C

Specifications

Color Rust
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: insoluble
Formula Weight 388.92
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material µ-Dichlorotetraethylene dirhodium(I)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0450108 Certificate TypeCertificate of Analysis Date14/02/2024 Catalog Number197591000,197590010
Lot NumberA0396502 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number197591000,197590010
Lot NumberA0372147 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number197591000,197590010
Lot NumberA0432197 Certificate TypeCertificate of Analysis Date26/07/2022 Catalog Number197591000,197590010
Lot NumberA0413479 Certificate TypeCertificate of Analysis Date31/10/2021 Catalog Number197591000,197590010
Product Identifier
  • µ-Dichlorotetraethylene dirhodium(I)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
μ-Dichlorotetraethylene dirhodium(I), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.