Learn More
Diclofenac diethylamine, MedChemExpress
MedChemExpress Diclofenac diethylamine is a potent and nonselective anti-inflammatory agent, acts as a COX inhibitor, with IC50s of 4 and 1.3 nM for human COX-1 and COX-2 in CHO cells, and 5.1 and 0.84 μM for ovine COX-1 and COX-2, respectively. Diclofenac diethylamine induces apoptosis of neural stem cells (NSCs) via the activation of the caspase cascade.
£77.44 - £127.05
Specifications
Chemical Name or Material | Diclofenac diethylamine |
---|---|
CAS | 78213-16-8 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C18H22Cl2N2O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16456298
|
Medchem Express
HY-15036A |
10 g |
£127.05
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16448887
|
Medchem Express
HY-15036A |
5 g |
£77.44
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Diclofenac diethylamine is a potent and nonselective anti-inflammatory agent, acts as a COX inhibitor, with IC50s of 4 and 1.3 nM for human COX-1 and COX-2 in CHO cells, and 5.1 and 0.84 μM for ovine COX-1 and COX-2, respectively. Diclofenac diethylamine induces apoptosis of neural stem cells (NSCs) via the activation of the caspase cascade.Specifications
Diclofenac diethylamine | |
Off-White | |
C18H22Cl2N2O2 | |
O=C(O)CC1=CC=CC=C1NC2=C(Cl)C=CC=C2Cl.CCNCC | |
369.29 | |
Research | |
COVID-19-immunoregulation |
78213-16-8 | |
Research | |
DMSO : 150 mg/mL (406.18 mM; Need ultrasonic) | |
369.29 | |
99.55% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.