missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diclofenac sodium salt, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Nonsteroidal, anti-inflammatory drug (NSAID); inhibits COX-1 and COX-2 (IC50 values are 0.075 μM for COX-1 and 0.038 μM for COX-2 in human whole blood assay).
Specifications
Specifications
CAS | 15307-79-6 |
Molecular Formula | C14H10Cl2NNaO2 |
Quantity | 50 mg |
Synonym | diclofenac sodium, sodium diclofenac, diclofenac sodium salt, voltaren, solaraze, anthraxiton, prophenatin, allvoran, batafil, delimon |
InChI Key | KPHWPUGNDIVLNH-UHFFFAOYSA-M |
SMILES | C1=CC=C(C(=C1)CC(=O)[O-])NC2=C(C=CC=C2Cl)Cl.[Na+] |
IUPAC Name | sodium;2-[2-(2,6-dichloroanilino)phenyl]acetate |
Molecular Weight (g/mol) | 318.129 |
PubChem CID | 5018304 |
ChEBI | CHEBI:4509 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction