missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicobalt octacarbonyl, 95%, stabilized, Thermo Scientific Chemicals
37.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 51.0°C to 52.0°C |
Color | Orange-Red to Red |
Density | 1.8100g/mL |
Flash Point | −13°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min |
Packaging | Glass bottle |
Linear Formula | Co2(CO)8 |
Quantity | 5 g |
Merck Index | 15,3095 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Dicobalt octacarbonyl, stabilized
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 2
- Respiratory sensitiser Category 1
- Self-heating substance Category 1
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H336-May cause drowsiness or dizziness.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H251-Self-heating: may catch fire.
- P235+P410-Keep cool. Protect from sunlight.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'