missing translation for 'onlineSavingsMsg'
Learn More

Dicumyl peroxide, 99%, Thermo Scientific Chemicals

Product Code. 10536821
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-43-3
Molecular Formula C18H22O2
Molecular Weight (g/mol) 270.37
MDL Number MFCD00036227
InChI Key XMNIXWIUMCBBBL-UHFFFAOYSA-N
Synonym dicumyl peroxide, cumene peroxide, cumyl peroxide, percumyl d, perkadox b, diisopropylbenzene peroxide, perkadox bc, perkadox sb, dicumenyl peroxide, peroxide, bis 1-methyl-1-phenylethyl
PubChem CID 6641
IUPAC Name 2-(2-phenylpropan-2-ylperoxy)propan-2-ylbenzene
SMILES CC(C)(C1=CC=CC=C1)OOC(C)(C)C2=CC=CC=C2

Specifications

Melting Point 38.0°C to 41.0°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Iodometry)
Packaging Plastic bottle
Linear Formula [C6H5C(CH3)2]2O2
Quantity 500 g
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 270.37
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material Dicumyl peroxide, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1A
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H242-Heating may cause a fire.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dicumyl peroxide, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.