missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicyandiamide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
500 g
2500 g
Description
Dicyandiamide is used in the synthesis of barbiturates. It is used as a stabilizer of ammonium dinitramide melt. It is used as hardener.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dicyandiamide is used in the synthesis of barbiturates. It is used as a stabilizer of ammonium dinitramide melt. It is used as hardener.
Solubility
Soluble in Water (32g/L ) at 20°C
Notes
Store at 2-8°C. Incompatible with strong acids, strong oxidizing agents, strong acids, strong bases, ammonium nitrate, potassium chlorates.
Dicyandiamide is used in the synthesis of barbiturates. It is used as a stabilizer of ammonium dinitramide melt. It is used as hardener.
Solubility
Soluble in Water (32g/L ) at 20°C
Notes
Store at 2-8°C. Incompatible with strong acids, strong oxidizing agents, strong acids, strong bases, ammonium nitrate, potassium chlorates.
Specifications
Specifications
Melting Point | 208°C to 211°C |
Density | 1.4 g/mL |
Assay Percent Range | 99% |
Quantity | 500 g |
Beilstein | 605637 |
Merck Index | 14,3092 |
Solubility Information | Soluble in Water (32g/L ) at 20°C |
Formula Weight | 84.08 |
Percent Purity | 99% |
Chemical Name or Material | Dicyandiamide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'