missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicyclopentadiene, 95%, stabilized with 100-200 ppm 4-tert-Butylcatechol, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spezifikation
Spezifikation
Melting Point | 11.0°C to 13.0°C |
Density | 0.9820g/mL |
Boiling Point | 170.0°C |
Flash Point | 32°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 91% min. (endo-Dicyclopentadiene) (GC) |
Packaging | Glass bottle |
Refractive Index | 1.5100 to 1.5120 |
Quantity | 1 L |
Beilstein | 05,II,391 |
Mehr anzeigen |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0462144 | Certificate of Analysis | 02/07/2024 | 150760100,150760025,150760010,150761000 |
A0444831 | Certificate of Analysis | 25/08/2023 | 150760100,150760025,150760010,150761000 |
A0452993 | Certificate of Analysis | 11/05/2023 | 150760100,150760025,150760010,150761000 |
A0451889 | Certificate of Analysis | 02/05/2023 | 150760100,150760025,150760010,150761000 |
A0422595 | Certificate of Analysis | 26/09/2022 | 150760100,150760025,150760010,150761000 |
Sicherheit und Handhabung



Produktidentifikator
- Dicyclopentadiene, stabilized
- Danger
- Acute toxicity Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- H335-May cause respiratory irritation.
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
- H330-Fatal if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- P310-Immediately call a POISON CENTER/doctor/.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Dicyclopentadiene, 95%, stabilized with 100-200 ppm 4-tert-Butylcatechol, Thermo Scientific Chemicals >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur