missing translation for 'onlineSavingsMsg'
Learn More
Dicyclopentadiene diepoxide, 98% Product Code.: 15361286

Dicyclopentadiene diepoxide, 98%

Product Code. 15361286
100 g
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy

Product Code. 15361286

Brand: Thermo Scientific Acros 348511000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 81-21-0 | C10H12O2 | 164.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 81-21-0
Melting Point 185.0°C to 189.0°C
Boiling Point 326.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Molecular Formula C10H12O2
Quantity 100 g
Synonym dicyclopentadiene dioxide, dicyclopentadiene diepoxide, unox epoxide 207, epoxide 207, bicyclopentadiene dioxide, unox 207x, unox 207, dicyclopentadiene dioxide van, 1,2:5,6-diepoxyhexahydro-4,7-methanoindan, 4,7-methanoindan, 1,2:5,6-diepoxyhexahydro
Solubility Information Solubility in water: 1.4g/L (20°C).
InChI Key BQQUFAMSJAKLNB-UHFFFAOYSA-N
SMILES C1C2C3CC4C(C3C1C5C2O5)O4
Molecular Weight (g/mol) 164.2
PubChem CID 6673
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material Dicyclopentadiene diepoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0371077 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number348511000,348510250,348510050
Lot NumberA0427547 Certificate TypeCertificate of Analysis Date05/09/2022 Catalog Number348511000,348510250,348510050
compliance-icons
Product Identifier
  • Dicyclopentadiene diepoxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dicyclopentadiene diepoxide, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.