missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopentanylmethyl acrylate, Thermo Scientific Chemicals

Product Code. 15462737
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 93962-84-6
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.312
MDL Number MFCD22380704
InChI Key PMPNHSZLJPXGCD-UHFFFAOYSA-N
Synonym octahydro-4,7-methano-1h-indenyl methyl acrylate, acmc-20mslm, tricyclo 5.2.1.0 2 ,? decan-3-ylmethyl prop-2-enoate
PubChem CID 22051805
SMILES C=CC(=O)OCC1CCC2C1C3CCC2C3

Spezifikation

Color Yellow
Quantity 5 g
Formula Weight 220.31
Physical Form Liquid
Chemical Name or Material Dicyclopentanylmethyl acrylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'