Learn More
Usbeck Laborgeraete™ Die-cast Zinc Burette Clamps
Click to view available options
For Use With (Equipment):
1 Burette
2 Burette
Unit Size:
Each
Specifications
Specifications
| Quantity | Each |
| Type | Burette Clamp |
| Material | Die Cast Zinc, Nickel Plated, Plastic Coated |
| Includes | roller grips |
| For Use With (Equipment) | 2 Burette |
| Size (Metric) Grip | 0 to 40 mm |