Learn More
Diesel; Multi-Parameter Certified Reference Material, UKAS ISO 17034 / ISO 17025 Certified, 500 mL, Paragon Scientific™
Multi-Parameter Certified Reference Materials are premium level, dual certified reference materials certified in accordance with both ISO 17025 and ISO 17034 under our UKAS accreditation. Fully dual certified in accordance with ISO 17025 and ISO 17034 under our accreditation by UKAS.
Brand: Paragon Scientific CRMU-MPGO
Additional Details : CAS Number : 68334-30-5 Weight : 0.90000kg Transport : UN number : 1202 Chem class : 3 Pack group : III
Description
- Certified for Viscosity, Density, Flash Point, Cloud Point, CFPP & Distillation using ASTM / IP and EN test method protocols
- Highest level of accreditation guarantee, providing the most credible certified data currently available world-wide
- Low levels of uncertainty achieves maximum accuracy of data
- Multiple test parameters provide a flexible and cost effective solution
- All data generated exclusively by UKAS ISO 17025 accredited laboratories
- Fully traceable to international standards
- 18 month shelf life
- 500mL inert glass packaging with tamper evident cap provides absolute assurance of sample integrity
- Available from stock for immediate dispatch
- Manufactured in the United Kingdom
Specifications
UN 1202 |
Safety and Handling
- Multi-Parameter Certified Reference Material, diesel
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Carcinogenicity Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H332-Harmful if inhaled.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P331-Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : diesel fuel
Nominal values quoted. Certified Values will vary from batch to batch.