missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diesel Range Organics Mix, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| n-Decane | 124-18-5 | 1000μg/mL |
| n-Docosane | 629-97-0 | 1000μg/mL |
| n-Dodecane | 112-40-3 | 1000μg/mL |
| n-Eicosane | 112-95-8 | 1000μg/mL |
| n-Hexacosane | 630-01-3 | 1000μg/mL |
| n-Hexadecane | 544-76-3 | 1000μg/mL |
| n-Octacosane | 630-02-4 | 1000μg/mL |
| n-Octadecane | 593-45-3 | 1000μg/mL |
| n-Tetracosane | 646-31-1 | 1000μg/mL |
| n-Tetradecane | 629-59-4 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | Diesel Range Organics Mix |
| Packaging | 2mL amber prescored glass ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| Methods | For US EPA Method 8015 |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Solvent or Matrix | Methylene Chloride |
| Quantity | 1 mL |
Safety and Handling
Product Identifier
- Diesel Range Organics Mix
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H351-Suspected of causing cancer.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : dichloromethane
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction