missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethoxymethane, 99.7%, Honeywell™
Click to view available options
Quantity:
2 L
2.5 L
Packaging:
Glass bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -50°C |
Color | Colorless |
Density | 0.831g/mL at 25°C |
Boiling Point | 255°C |
Packaging | Glass bottle |
Linear Formula | CH2(OC2H5)2 |
Quantity | 2 L |
UN Number | UN2373 |
Beilstein | 1697253 |
Vapor Density | 3.6 (vs air) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'