Learn More
Diethoxymethylphenylsilane, 97+%, Thermo Scientific Chemicals
£28.90 - £85.50
Chemical Identifiers
CAS | 775-56-4 |
---|---|
Molecular Formula | C11H18O2Si |
Molecular Weight (g/mol) | 210.35 |
MDL Number | MFCD00048541 |
InChI Key | MNFGEHQPOWJJBH-UHFFFAOYSA-N |
Synonym | methylphenyldiethoxysilane, diethoxy methyl phenylsilane, diethoxy methyl phenyl silane, silane, diethoxymethylphenyl, benzene, diethoxymethylsilyl, pubchem10811, acmc-209pap, methyl-phenyl-diethoxysilane, diethoxy-methyl-phenyl-silane |
PubChem CID | 69891 |
IUPAC Name | diethoxy-methyl-phenylsilane |
SMILES | CCO[Si](C)(OCC)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11400098
|
Thermo Scientific Alfa Aesar
B23684.09 |
10 g |
£28.90
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410098
|
Thermo Scientific Alfa Aesar
B23684.18 |
50 g |
£85.50
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
775-56-4 | |
210.35 | |
MNFGEHQPOWJJBH-UHFFFAOYSA-N | |
69891 | |
CCO[Si](C)(OCC)C1=CC=CC=C1 |
C11H18O2Si | |
MFCD00048541 | |
methylphenyldiethoxysilane, diethoxy methyl phenylsilane, diethoxy methyl phenyl silane, silane, diethoxymethylphenyl, benzene, diethoxymethylsilyl, pubchem10811, acmc-209pap, methyl-phenyl-diethoxysilane, diethoxy-methyl-phenyl-silane | |
diethoxy-methyl-phenylsilane |
Specifications
117°C to 118°C (30mmHg) | |
C11H18O2Si | |
2938396 | |
methylphenyldiethoxysilane, diethoxy methyl phenylsilane, diethoxy methyl phenyl silane, silane, diethoxymethylphenyl, benzene, diethoxymethylsilyl, pubchem10811, acmc-209pap, methyl-phenyl-diethoxysilane, diethoxy-methyl-phenyl-silane | |
CCO[Si](C)(OCC)C1=CC=CC=C1 | |
210.35 | |
210.35 | |
1.4696 | |
0.963 |
775-56-4 | |
MFCD00048541 | |
88°C (190°F) | |
MNFGEHQPOWJJBH-UHFFFAOYSA-N | |
diethoxy-methyl-phenylsilane | |
69891 | |
≥97% | |
Moisture sensitive | |
Diethoxymethylphenylsilane |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
missing translation for 'einecsNumber' : 212-275-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only