missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 1,1-cyclobutanedicarboxylate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Diethyl 1,1-cyclobutanedicarboxylate is used as a pharmaceutical intermediate. Used as intermediate in the production of Carboplatin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethyl 1,1-cyclobutanedicarboxylate is used as a pharmaceutical intermediate. Used as intermediate in the production of Carboplatin.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Diethyl 1,1-cyclobutanedicarboxylate is used as a pharmaceutical intermediate. Used as intermediate in the production of Carboplatin.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
Density | 1.05 |
Boiling Point | 96°C to 98°C (8 mmHg) |
Flash Point | 89°C (192°F) |
Refractive Index | 1.436 |
Quantity | 10 g |
Beilstein | 2050195 |
Solubility Information | Insoluble in water. |
Formula Weight | 200.24 |
Percent Purity | 95% |
Chemical Name or Material | Diethyl 1,1-cyclobutanedicarboxylate |
RUO – Research Use Only
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto