missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1,4-dihydro-2,6-dimethylpyridine-3,5-dicarboxylate, 98%, Thermo Scientific Chemicals

Product Code. 15412276
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate is a hydrogen source in conjugate reduction and organocatalytic reductive amination. It is used as a hydrogen source in organocatalytic reductive amination and conjugate reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate is a hydrogen source in conjugate reduction and organocatalytic reductive amination. It is used as a hydrogen source in organocatalytic reductive amination and conjugate reduction.

Solubility
Soluble in organic solvents.

Notes
Desiccate at 4°C. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1149-23-1
Molecular Formula C13H19NO4
Molecular Weight (g/mol) 253.298
MDL Number MFCD00005951
InChI Key LJXTYJXBORAIHX-UHFFFAOYSA-N
Synonym diludine, diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate, hantzsch ester, ethidine, etidin, diethone, hantzsch's dihydropyridine, diethyl 2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate, diludin, 3,5-pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-, diethyl ester
PubChem CID 70849
IUPAC Name diethyl 2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C(C1)C(=O)OCC)C)C

Specifications

Melting Point 178°C to 183°C
Quantity 250 mg
Solubility Information Soluble in organic solvents.
Formula Weight 253.3
Percent Purity 98%
Chemical Name or Material Diethyl 1,4-dihydro-2,6-dimethylpyridine-3,5-dicarboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'