missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2,2-diethoxyethylphosphonate, 96%, Thermo Scientific Chemicals

Product Code. 11499693
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Diethyl 2,2-diethoxyethylphosphonate is used reactant for synthesis of α,β-alkenal derivatives by two-carbon homologation, lower rim-phosphonylated rexorcinol calix[4]arenes by condensation reactions, α-Phosphovinyl radicals via a radical trapping sequence, inhibitors of reverse transcriptase via 1,3-dipolar cycloadditions and for Friedel-Crafts reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 2,2-diethoxyethylphosphonate is used reactant for synthesis of α,β-alkenal derivatives by two-carbon homologation, lower rim-phosphonylated rexorcinol calix[4]arenes by condensation reactions, α-Phosphovinyl radicals via a radical trapping sequence, inhibitors of reverse transcriptase via 1,3-dipolar cycloadditions and for Friedel-Crafts reactions.

Solubility
Soluble in chloroform and dichloromethane. Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. It is hygroscopic in nature. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7598-61-0
Molecular Formula C10H23O5P
Molecular Weight (g/mol) 254.263
MDL Number MFCD00009236
InChI Key LUQYELQXRPNKRY-UHFFFAOYSA-N
Synonym diethyl 2,2-diethoxyethylphosphonate, diethyl 2,2-diethoxyethyl phosphonate, diethyl2,2-diethoxyethylphosphonate, 2,2-diethoxyethylphosphonic acid diethyl ester, phosphonic acid, 2,2-diethoxyethyl-, diethyl ester, 2,2-diethoxyethyl diethoxyphosphino-1-one, diethyl phosphonacetaldehyde diethyl acetal, diethylphosphonoacetaldehyde diethylacetal, 2-diethoxyphosphoryl-1,1-diethoxy-ethane
PubChem CID 82071
IUPAC Name 2-diethoxyphosphoryl-1,1-diethoxyethane
SMILES CCOC(CP(=O)(OCC)OCC)OCC

Specifications

Density 1.052
Boiling Point 101°C to 103°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4278
Quantity 10 g
Beilstein 1785614
Sensitivity Hygroscopic
Solubility Information Soluble in chloroform and dichloromethane. Not miscible or difficult to mix in water.
Formula Weight 254.26
Percent Purity 96%
Chemical Name or Material Diethyl 2,2-diethoxyethylphosphonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'