missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Diethyl 2,4-dimethylpyrrole-3,5-dicarboxylate, 97%

Product Code. 11479317
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 2436-79-5
Molecular Formula C12H17NO4
Molecular Weight (g/mol) 239.271
MDL Number MFCD00005218
InChI Key XSBSXJAYEPDGSF-UHFFFAOYSA-N
PubChem CID 75526
IUPAC Name diethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C1C)C(=O)OCC)C

Specifications

Melting Point 135°C to 138°C
Quantity 5 g
Synonym diethyl 2,4-dimethylpyrrole-3,5-dicarboxylate, 3,5-dimethyl-1h-pyrrole-2,4-dicarboxylic acid diethyl ester, knorr's pyrrole, 2,4-diethyl 3,5-dimethyl-1h-pyrrole-2,4-dicarboxylate, 1h-pyrrole-2,4-dicarboxylic acid, 3,5-dimethyl-, diethyl ester, 2,4-dimethyl-3,5-dicarbethoxypyrrole, diethyl 2,4-dimethyl-1h-pyrrole-3,5-dicarboxylate, diethyl2,4-dimethylpyrrole-3,5-dicarboxylate, pyrrole-2,4-dicarboxylic acid, 3,5-dimethyl-, diethyl ester, diethyl 2 ,4-dimethylpyrrole-3,5-dicarboxylate
Formula Weight 239.27
Percent Purity 97%
Chemical Name or Material Diethyl 2,4-dimethylpyrrole-3,5-dicarboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'