missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 2-bromoethylphosphonate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
It is employed as a reactant involved in radical coupling, synthesis of organosoluble zirconium phosphonate nanocomposites, click-chemistry, asymmetric epoxidation of unfunctionalized olefins, Negishi alkyl-aryl cross-coupling, and in synthesis of hydrolytically stable phosphonic acids for use as dental adhesives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a reactant involved in radical coupling, synthesis of organosoluble zirconium phosphonate nanocomposites, click-chemistry, asymmetric epoxidation of unfunctionalized olefins, Negishi alkyl-aryl cross-coupling, and in synthesis of hydrolytically stable phosphonic acids for use as dental adhesives.
Solubility
Soluble in water(20 g/L) (25°C).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is employed as a reactant involved in radical coupling, synthesis of organosoluble zirconium phosphonate nanocomposites, click-chemistry, asymmetric epoxidation of unfunctionalized olefins, Negishi alkyl-aryl cross-coupling, and in synthesis of hydrolytically stable phosphonic acids for use as dental adhesives.
Solubility
Soluble in water(20 g/L) (25°C).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.275 |
Boiling Point | 98°C to 100°C (1 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.4605 |
Quantity | 5 g |
Beilstein | 1766077 |
Solubility Information | Soluble in water(20g/L) (25°C). |
Formula Weight | 245.06 |
Percent Purity | 97% |
Chemical Name or Material | Diethyl 2-bromoethylphosphonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'