missing translation for 'onlineSavingsMsg'
Learn More
Diethyl 3,4-furandicarboxylate, 95% Product Code.: 10658824

Diethyl 3,4-furandicarboxylate, 95%

Product Code. 10658824
5 g, Glass bottle
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10658824

Brand: Thermo Scientific Acros 156510050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 30614-77-8 | C10H12O5 | 212.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 30614-77-8
Color Colorless
Density 1.1400g/mL
Boiling Point 155°C (13.0 mmHg)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C10H12O5
Refractive Index 1.4715 to 1.4735
MDL Number MFCD00005349
Quantity 5 g
Specific Gravity 1.14
Synonym diethyl 3,4-furandicarboxylate, 3,4-furandicarboxylic acid, diethyl ester, 3,4-diethyl furan-3,4-dicarboxylate, furan-3,4-dicarboxylic acid diethyl ester, 5-18-07-00053 beilstein handbook reference, zodfwnhyqarjlc-uhfffaoysa, 3,4-furandicarboxylicacid, 3,4-diethyl ester
InChI Key ZODFWNHYQARJLC-UHFFFAOYSA-N
SMILES CCOC(=O)C1=COC=C1C(=O)OCC
IUPAC Name diethyl furan-3,4-dicarboxylate
Molecular Weight (g/mol) 212.20
PubChem CID 35399
Formula Weight 212.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diethyl 3, 4-furandicarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl 3,4-furandicarboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyl 3,4-furandicarboxylate, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.