missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 4-aminobenzylphosphonate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
Specifications
Specifications
Melting Point | 90°C to 94°C |
Color | Beige to Brown |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Linear Formula | (C2H5O)2P(O)CH2C6H4NH2 |
Quantity | 5 g |
Formula Weight | 243.24 |
Percent Purity | 98% |
Physical Form | Crystalline Powder or Crystals |
Show More |
Safety and Handling
Product Identifier
- Diethyl 4-aminobenzylphosphonate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction