missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (4-nitrobenzyl)phosphonate, 98%, Thermo Scientific Chemicals

Product Code. 10662012
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2609-49-6
Molecular Formula C11H16NO5P
Molecular Weight (g/mol) 273.22
MDL Number MFCD00047747
InChI Key FORMFFDDQMCTCT-UHFFFAOYSA-N
PubChem CID 256065
IUPAC Name 1-(diethoxyphosphorylmethyl)-4-nitrobenzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)[N+](=O)[O-])OCC

Specifications

Color Orange-Yellow to Red
Density 1.2400g/mL
Boiling Point 151°C (0.1 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5220 to 1.526
Quantity 5 g
Beilstein 16, IV, 1096
Specific Gravity 1.24
Synonym diethyl 4-nitrobenzylphosphonate, diethyl 4-nitrobenzyl phosphonate, diethyl 4-nitrophenyl methylphosphonate, 1-diethoxyphosphorylmethyl-4-nitro-benzene, 1-diethoxyphosphorylmethyl-4-nitrobenzene, phosphonic acid, 4-nitrophenyl methyl-, diethyl ester, phosphonic acid, p-4-nitrophenyl methyl-, diethyl ester, zlchem 660, diethyl4-nitrobenzylphosphonate
Formula Weight 273.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diethyl (4-nitrobenzyl)phosphonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.