missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl bis(hydroxymethyl)malonate, 95%, Thermo Scientific Chemicals
£41.50 - £776.00
Chemical Identifiers
CAS | 20605-01-0 |
---|---|
Molecular Formula | C9H16O6 |
Molecular Weight (g/mol) | 220.22 |
MDL Number | MFCD00009130 |
InChI Key | WIOHBOKEUIHYIC-UHFFFAOYSA-N |
Synonym | diethyl bis hydroxymethyl malonate, diethyl 2,2-bis hydroxymethyl malonate, propanedioic acid, bis hydroxymethyl-, diethyl ester, 1,3-diethyl 2,2-bis hydroxymethyl propanedioate, diethyl 2,2-bis hydroxymethyl propanedioate, 2,2-bis hydroxymethyl malonic acid diethyl ester, bis hydroxymethyl malonic acid diethyl ester, diethyl 2,2-bis hydroxymethyl propane-1,3-dioate, acmc-20al8x, ksc490a8h |
PubChem CID | 311844 |
IUPAC Name | diethyl 2,2-bis(hydroxymethyl)propanedioate |
SMILES | CCOC(=O)C(CO)(CO)C(=O)OCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10427760
|
Thermo Scientific Acros
189530250 |
25 g | Glass bottle |
£41.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10266770
|
Thermo Scientific Acros
189531000 |
100 g | Glass bottle |
£135.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10036353
|
Thermo Scientific Acros
189535000 |
500 g | Glass Bottle |
£776.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20605-01-0 | |
220.22 | |
WIOHBOKEUIHYIC-UHFFFAOYSA-N | |
311844 | |
CCOC(=O)C(CO)(CO)C(=O)OCC |
C9H16O6 | |
MFCD00009130 | |
diethyl bis hydroxymethyl malonate, diethyl 2,2-bis hydroxymethyl malonate, propanedioic acid, bis hydroxymethyl-, diethyl ester, 1,3-diethyl 2,2-bis hydroxymethyl propanedioate, diethyl 2,2-bis hydroxymethyl propanedioate, 2,2-bis hydroxymethyl malonic acid diethyl ester, bis hydroxymethyl malonic acid diethyl ester, diethyl 2,2-bis hydroxymethyl propane-1,3-dioate, acmc-20al8x, ksc490a8h | |
diethyl 2,2-bis(hydroxymethyl)propanedioate |
Specifications
20605-01-0 | |
White | |
Authentic | |
C9H16O6 | |
MFCD00009130 | |
WIOHBOKEUIHYIC-UHFFFAOYSA-N | |
diethyl 2,2-bis(hydroxymethyl)propanedioate | |
311844 | |
95% | |
Diethyl bis(hydroxymethyl)malonate |
45°C to 52°C | |
>110°C | |
95% | |
(HOCH2)2C(CO2C2H5)2 | |
diethyl bis hydroxymethyl malonate, diethyl 2,2-bis hydroxymethyl malonate, propanedioic acid, bis hydroxymethyl-, diethyl ester, 1,3-diethyl 2,2-bis hydroxymethyl propanedioate, diethyl 2,2-bis hydroxymethyl propanedioate, 2,2-bis hydroxymethyl malonic acid diethyl ester, bis hydroxymethyl malonic acid diethyl ester, diethyl 2,2-bis hydroxymethyl propane-1,3-dioate, acmc-20al8x, ksc490a8h | |
CCOC(=O)C(CO)(CO)C(=O)OCC | |
220.22 | |
220.22 | |
Crystalline Solid |
Safety and Handling
GHS H Statement
Causes skin irritation.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 243-916-8
RUO – Research Use Only