missing translation for 'onlineSavingsMsg'
Learn More

Diethyl chlorophosphite, 90%, tech., Thermo Scientific Chemicals

Product Code. 10264040
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-57-1
Molecular Formula C4H10ClO2P
Molecular Weight (g/mol) 156.55
MDL Number MFCD00009074
InChI Key TXHWYSOQHNMOOU-UHFFFAOYSA-N
Synonym diethyl chlorophosphite, ethyl phosphorochloridite, diethyl phosphorochloridite, chlorodiethoxyphosphine, phosphorochloridous acid, diethyl ester, unii-f414s3v8k6, diethyl chlorophosphonite, chloro diethoxy phosphane, diethychlorophosphite, ethyl chlorophosphite
PubChem CID 68530
IUPAC Name chloro(diethoxy)phosphane
SMILES CCOP(OCC)Cl

Specifications

Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 153.0°C to 155.0°C
Flash Point <1°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (Argentometry)
Packaging Glass bottle
Linear Formula (C2H5O)2PCl
Refractive Index 1.4380 to 1.4420
Quantity 25 mL
Fieser 01,248
Merck Index 15, 3144
Specific Gravity 1.09
Solubility Information Solubility in water: reacts
Formula Weight 156.55
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Diethyl chlorophosphite, tech., 90%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl chlorophosphite
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'